Trifluoroacetic acid
Catalog # APOSPC8841-1KG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:114.02 g/mol
Boiling Pt:72.4 °C
Melting Pt:15.4 °C
Density:1.489 g/cm³ (25 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:200-929-3
UN:2699
ADR:8,I
:GHS05
:GHS07
CAS Number:76-05-1
Formula:C₂HF₃O₂
MDL Number:MFCD00004169