
2,6-Dinitro-4-(trifluoromethyl)aniline 97%
Catalog # APOSPC8461-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:251.12 g/mol
Melting Pt:144…149 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:445-66-9
Formula:C₇H₄F₃N₃O₄
MDL Number:MFCD00024260