Order Entry
Germany
ContactUsLinkComponent
Diethyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]malonate 97%
Diethyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]malonate 97%
Catalog # APOSPC8453-1G
Supplier:  Apollo Scientific
CAS Number:  
Diethyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]malonate 97%
Catalog # APOSPC8453-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :339.7 g/mol
Boiling Pt:120…124 °C/0.5mmHg
Melting Pt:43…45 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:172527-71-8
Formula:C₁₃H₁₃ClF₃NO₄
MDL Number:MFCD00231883