N-(Furan-2-ylmethyl) 2-bromo-5-fluorobenzamide
Catalog # APOSPC7978-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:923722-86-5
Formula:C₁₂H₉BrFNO₂
MDL Number:MFCD08872270