Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-5-fluoro-N-methylbenzamide
2-Bromo-5-fluoro-N-methylbenzamide 98%
2-Bromo-5-fluoro-N-methylbenzamide 98%
2-Bromo-5-fluoro-N-methylbenzamide 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Bromo-5-fluoro-N-methylbenzamide 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
171426-13-4
:
Formula:
C₈H₇BrFNO
MDL Number:
MFCD09475889
Storage Temperature:
Ambient