Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dimethyl-4-fluoronitrobenzene
2,3-Dimethyl-4-fluoronitrobenzene
2,3-Dimethyl-4-fluoronitrobenzene
Irritant
2,3-Dimethyl-4-fluoronitrobenzene
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,3-Dimethyl-4-fluoronitrobenzene
Supplier:
Apollo Scientific
About this item
CAS Number:
1736-87-4
:
Formula:
C₈H₈FNO₂
MDL Number:
MFCD05663712
:
169.16 g/mol
:
GHS07
Storage Temperature:
Ambient