
Ethyl (2,3,4,5-tetrafluorobenzoyl)acetate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:94695-50-8
:
Formula:C₁₁H₈F₄O₃
MDL Number:MFCD00523020
:264.18 g/mol
Boiling Pt:100 °C/0.075mmHg
Melting Pt:48 °C
:GHS07
Storage Temperature:Ambient