Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 2,4,5-trifluorobenzoate
Ethyl-2,4,5-trifluorobenzoate
Ethyl-2,4,5-trifluorobenzoate
Irritant
Ethyl-2,4,5-trifluorobenzoate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Ethyl-2,4,5-trifluorobenzoate
Supplier:
Apollo Scientific
About this item
CAS Number:
351354-41-1
:
Formula:
C₉H₇F₃O₂
MDL Number:
MFCD03094444
:
204.1459 g/mol
Boiling Pt:
206 °C
:
GHS07
Storage Temperature:
Ambient