
2-(Trifluoromethyl)phenyl isothiocyanate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1743-86-8
:
Formula:C₈H₄F₃NS
MDL Number:MFCD00039644
:203.19 g/mol
Boiling Pt:95…96 °C/10mmHg
Density:1.346 g/cm³ (25 °C)
Flash Pt:105 °C
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS07