Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Trifluoromethyl)nicotinic acid
4-(Trifluoromethyl)nicotinic acid 97%
4-(Trifluoromethyl)nicotinic acid 97%
4-(Trifluoromethyl)nicotinic acid 97%
Supplier:
Apollo Scientific
Certificates
Expand image
4-(Trifluoromethyl)nicotinic acid 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
158063-66-2
:
Formula:
C₇H₄F₃NO₂
MDL Number:
MFCD00082626
:
191.11 g/mol
Melting Pt:
146…149 °C
:
GHS07
Storage Temperature:
Ambient