1-Methyl-4-[2-nitro-4-(trifluoromethyl)phenyl]piperazine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:298703-18-1
:
Formula:C₁₂H₁₅ClF₃N₃O₂
MDL Number:MFCD01208354
:325.72 g/mol
Melting Pt:200…210 °C
Storage Temperature:Ambient