Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,4-Difluorocyclohexylamine hydrochloride
4,4-Difluorocyclohexylamine hydrochloride 98%
4,4-Difluorocyclohexylamine hydrochloride 98%
Irritant
4,4-Difluorocyclohexylamine hydrochloride 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4,4-Difluorocyclohexylamine hydrochloride 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
675112-70-6
:
Formula:
C₆H₁₂ClF₂N
MDL Number:
MFCD06246881
:
171.62 g/mol
Melting Pt:
106…107 °C
:
GHS07
Storage Temperature:
Ambient