Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4,5-Trifluorobenzoic acid
2,4,5-Trifluorobenzoic acid 99%
2,4,5-Trifluorobenzoic acid 99%
Irritant
2,4,5-Trifluorobenzoic acid 99%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,4,5-Trifluorobenzoic acid 99%
Supplier:
Apollo Scientific
About this item
CAS Number:
446-17-3
:
Formula:
C₇H₃F₃O₂
MDL Number:
MFCD00013306
:
176.09 g/mol
Melting Pt:
98…99 °C
:
GHS07
Storage Temperature:
Ambient