Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Trifluoroacetic acid
Trifluoroacetic acid 99,9%, Biochemical Grade
Trifluoroacetic acid 99,9%, Biochemical Grade
:
Apollo Scientific
Trifluoroacetic acid 99,9%, Biochemical Grade
:
Apollo Scientific
:
76-05-1
:
:
C₂HF₃O₂
:
MFCD00004169
:
114.02 g/mol
:
72.4 °C
:
–15.4 °C
:
1.48 g/cm³ (25 °C)
:
>100 °C
:
Ambient
:
200-929-3
:
2699
:
8,I
:
GHS05
:
GHS07