

2,3,5,6-Tetrafluorotoluene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5230-78-4
:
Formula:C₇H₄F₄
:164.1 g/mol
Boiling Pt:125…126 °C
Density:1.347 g/cm³ (25 °C)
Flash Pt:25 °C
Storage Temperature:Ambient
EINECS:226-028-5
UN:1993
ADR:3,III
:GHS02
:GHS07