

2,3,5,6-Tetrafluorothiophenol 98%
Catalog # APOSPC6920-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:182.14 g/mol
Boiling Pt:152…153 °C
Density:1.53 g/cm³ (25 °C)
Flash Pt:48 °C
Storage Temperature:Refrigerator
EINECS:212-210-1
UN:3336
ADR:3,III
:GHS02
:GHS07
CAS Number:769-40-4
Formula:C₆H₂F₄S
MDL Number:MFCD00004829