
4-Methoxymethyl-2,3,5,6-tetrafluorobenzenemethanol 97%
Catalog # APOSPC6638-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:224.16 g/mol
Melting Pt:45…50 °C
Flash Pt:107 °C
:GHS07
Storage Temperature:Ambient
CAS Number:83282-91-1
Formula:C₉H₈F₄O₂
MDL Number:MFCD08688060