Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium hexafluorosilicate
Sodium hexafluorosilicate 98%
Sodium hexafluorosilicate 98%
APOSPC6511-100G
:
Apollo Scientific
:
Sodium hexafluorosilicate 98%
APOSPC6511-100G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 g
Seq:
3401
:
:
188.06 g/mol
:
2.679 g/cm³ (25 °C)
:
Ambient
:
240-934-8
:
2674
:
6.1,III
:
GHS06
:
16893-85-9
:
F₆Na₂Si
:
MFCD00003491