Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Fluoro-3-(trifluoromethoxy)phenylboronic acid
2-Fluoro-3-(trifluoromethoxy)phenylboronic acid
2-Fluoro-3-(trifluoromethoxy)phenylboronic acid
2-Fluoro-3-(trifluoromethoxy)phenylboronic acid
Supplier:
Apollo Scientific
Certificates
Expand image
2-Fluoro-3-(trifluoromethoxy)phenylboronic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
881402-25-1
:
Formula:
C₇H₅BF₄O₃
MDL Number:
MFCD11045053
:
223.917 g/mol
:
GHS07
Storage Temperature:
Ambient