Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4,5-Trifluoropyrimidine
2,4,5-Trifluoropyrimidine
2,4,5-Trifluoropyrimidine
:
Apollo Scientific
2,4,5-Trifluoropyrimidine
:
Apollo Scientific
:
17573-79-4
:
:
C₄HF₃N₂
:
MFCD11845791
:
88…91 °C/23mmHg
:
2810
:
6.1,III
:
134.06 g/mol
:
GHS06
:
Ambient