Order Entry
Germany
ContactUsLinkComponent
4-(Bromomethyl)-2,3,5,6-tetrafluorobenzotrifluoride
Corrosive
4-(Bromomethyl)-2,3,5,6-tetrafluorobenzotrifluoride
Catalog # APOSPC5749-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
4-(Bromomethyl)-2,3,5,6-tetrafluorobenzotrifluoride
Catalog # APOSPC5749-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :311 g/mol
Density:1.864 g/cm³ (25 °C)
Flash Pt:99 °C
Storage Temperature:Refrigerator
UN:3265
ADR:8,II
 :GHS05
CAS Number:76437-40-6
Formula:C₈H₂BrF₇
MDL Number:MFCD00191855