Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4,5-Trifluorobenzenesulphonamide
2,4,5-Trifluorobenzenesulphonamide
2,4,5-Trifluorobenzenesulphonamide
2,4,5-Trifluorobenzenesulphonamide
Supplier:
Apollo Scientific
Certificates
Expand image
2,4,5-Trifluorobenzenesulphonamide
Supplier:
Apollo Scientific
About this item
CAS Number:
287172-63-8
:
Formula:
C₆H₄F₃NO₂S
MDL Number:
MFCD01569462
:
211.16 g/mol
Storage Temperature:
Ambient