

Di-tert-butyl(methyl)phosphonium tetrafluoroborate 95%
Catalog # APOSPC56018-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:248.05 g/mol
Storage Temperature:Ambient
EINECS:808-098-4
UN:1759
ADR:8,III
:GHS05
:GHS07
CAS Number:479094-62-7
:870777-30-3
Formula:C₉H₂₂BF₄P
MDL Number:MFCD03840579