(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid 97%
Catalog # APOSPC520853-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:136.10 g/mol
Storage Temperature:Ambient
CAS Number:128073-33-6
Formula:C₅H₆F₂O₂
MDL Number:MFCD07777163