Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4,5-Trifluoro-3-methoxybenzoic acid
2,4,5-Trifluoro-3-methoxybenzoic acid 98%
2,4,5-Trifluoro-3-methoxybenzoic acid 98%
:
Apollo Scientific
2,4,5-Trifluoro-3-methoxybenzoic acid 98%
:
Apollo Scientific
:
112811-65-1
:
C₈H₅F₃O₃
:
MFCD00153201
:
206.12 g/mol
:
105…119 °C
:
GHS07
:
Ambient