Order Entry
Germany
ContactUsLinkComponent
(2R,3S,4R,5R)-5-((Benzoyloxy)methyl)-3-fluorotetrahydrofuran-2,4-diyl dibenzoate 98%
Irritant
(2R,3S,4R,5R)-5-((Benzoyloxy)methyl)-3-fluorotetrahydrofuran-2,4-diyl dibenzoate 98%
Catalog # APOSPC49254-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(2R,3S,4R,5R)-5-((Benzoyloxy)methyl)-3-fluorotetrahydrofuran-2,4-diyl dibenzoate 98%
Catalog # APOSPC49254-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :464.44 g/mol
Melting Pt:76…78 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:97614-43-2
Formula:C₂₆H₂₁FO₇
MDL Number:MFCD00083339