Order Entry
Germany
ContactUsLinkComponent
4-Hydroxy-6-(trifluoromethyl)pyrimidine 97%
Irritant
4-Hydroxy-6-(trifluoromethyl)pyrimidine 97%
Catalog # APOSPC49210-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Hydroxy-6-(trifluoromethyl)pyrimidine 97%
Catalog # APOSPC49210-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :164.09 g/mol
Melting Pt:169…171 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1546-78-7
Formula:C₅H₃F₃N₂O
MDL Number:MFCD00192530