
2,6-Difluorobenzyl mercaptan
Catalog # APOSPC48785-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:GHS07
Storage Temperature:Refrigerator
CAS Number:878204-82-1
Formula:C₇H₆F₂S
MDL Number:MFCD06798019