Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Hydroxy-3-(trifluoromethyl)pyridine
2-Hydroxy-3-(trifluoromethyl)pyridine 97%
2-Hydroxy-3-(trifluoromethyl)pyridine 97%
Irritant
2-Hydroxy-3-(trifluoromethyl)pyridine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Hydroxy-3-(trifluoromethyl)pyridine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
22245-83-6
:
Formula:
C₆H₄F₃NO
MDL Number:
MFCD00153194
:
163.1 g/mol
Melting Pt:
150…155 °C
:
GHS07
Storage Temperature:
Ambient