
Hexafluorosilicic acid 23% in aqueous solution
Catalog # APOSPC4797-1KG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:144.09 g/mol
Melting Pt:15…17 °C
Density:1.22 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:241-034-8
UN:1778
ADR:8,II
:GHS05
CAS Number:16961-83-4
Formula:H₂F₆Si
MDL Number:MFCD00036289