Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Cyclopropyl-4-fluorobenzylamine
N-Cyclopropyl-4-fluorobenzylamine
N-Cyclopropyl-4-fluorobenzylamine
APOSPC449006-5G
:
Apollo Scientific
:
N-Cyclopropyl-4-fluorobenzylamine
APOSPC449006-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
165.211 g/mol
:
Ambient
:
625437-46-9
:
C₁₀H₁₂FN
:
MFCD07330007