N'-[1-Amino-1-(3-chloro-4-fluorophenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1053655-93-8
:
Formula:C₁₂H₁₅ClFN₃O₂
MDL Number:MFCD10568190
:287.72 g/mol
Storage Temperature:Ambient