Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Fluoro-m-toluidine
6-Fluoro-m-toluidine
6-Fluoro-m-toluidine
:
Apollo Scientific
6-Fluoro-m-toluidine
:
Apollo Scientific
:
452-84-6
:
:
C₇H₈FN
:
MFCD00007654
:
125.15 g/mol
:
80…86 °C/11mmHg
:
1.109 g/cm³ (25 °C)
:
81 °C
:
GHS07
:
Ambient