1H,1H-Heptafluorobutyl acrylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:424-64-6
:
Formula:C₇H₅F₇O₂
MDL Number:MFCD00039252
:254.11 g/mol
Boiling Pt:120…122 °C/743mmHg
Density:1.418 g/cm³ (25 °C)
Flash Pt:31 °C
Storage Temperature:Ambient
EINECS:207-036-8
UN:3272
ADR:3,III
:GHS02
:GHS07