

1-Chloro-2,3,5,6-tetrafluorobenzene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1835-61-6
:
Formula:C₆HClF₄
MDL Number:MFCD00012237
:184.5200 g/mol
Boiling Pt:125…126.5 °C/738mmHg
Density:1.538 g/cm³ (25 °C)
Flash Pt:43 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07