Order Entry
Germany
ContactUsLinkComponent
N-Succinimidyl trifluoroacetate 95%
Corrosive
Irritant
N-Succinimidyl trifluoroacetate 95%
Catalog # APOSPC430592-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
N-Succinimidyl trifluoroacetate 95%
Catalog # APOSPC430592-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :211.10 g/mol
Melting Pt:176 °C (dec.)
Storage Temperature:Ambient
UN:3261
ADR:8,III
 :GHS05
 :GHS07
CAS Number:5672-89-9
Formula:C₆H₄F₃NO₄
MDL Number:MFCD00595252