2-Chloro-4-fluoro-5-sulphamoylbenzoic acid
Catalog # APOSPC430506-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:253.6353 g/mol
Storage Temperature:Ambient
CAS Number:4793-24-2
Formula:C₇H₅ClFNO₄S
MDL Number:MFCD01321357