tert-Butyl-trans-3-(tert-butoxycarbonyloxy)-4-fluoropyrrolidine-1-carboxylate
Catalog # APOSPC430191-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:305.35 g/mol
Storage Temperature:Ambient
CAS Number:1373503-73-1
Formula:C₁₄H₂₄FNO₅
MDL Number:MFCD22199429