cis-1-Boc-3-fluoro-4-hydroxypiperidine
Catalog # APOSPC430153-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:219.26 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:955028-88-3
Formula:C₁₀H₁₈FNO₃
MDL Number:MFCD18791209