1-(2,2-Difluoroethyl)-1H-pyrazol-4-amine hydrochloride
Catalog # APOSPC410084-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:183.59 g/mol
Storage Temperature:Ambient
CAS Number:1197231-23-4
Formula:C₅H₈ClF₂N₃
MDL Number:MFCD15142911