Order Entry
Germany
ContactUsLinkComponent
3-(4-Fluorophenyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine 95+%
Irritant
3-(4-Fluorophenyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine 95+%
Catalog # APOSPC409516-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-(4-Fluorophenyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine 95+%
Catalog # APOSPC409516-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :217.25 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:916423-52-4
Formula:C₁₂H₁₂FN₃
MDL Number:MFCD08460453