3-(2-Fluorophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine 95+%
Catalog # APOSPC409514-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:217.25 g/mol
Storage Temperature:Ambient
CAS Number:916423-12-6
Formula:C₁₂H₁₂FN₃
MDL Number:MFCD09258791