N-Piperidin-4-yl-2-(trifluoromethyl)-1,6-naphthyridin-5-amine hydrochloride monohydrate 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1452488-43-5
:
Formula:C₁₄H₁₈ClF₃N₄O
MDL Number:MFCD08567943
:350.77 g/mol
Storage Temperature:Ambient