6-(Trifluormethyl)nicotinonitrile 98%
Catalog # APOSPC3838-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:172.1100 g/mol
Melting Pt:37…41 °C
Flash Pt:82.2 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,II
:GHS05
:GHS06
CAS Number:216431-85-5
Formula:C₇H₃F₃N₂
MDL Number:MFCD03001347