Order Entry
Germany
ContactUsLinkComponent
4'-Fluoro-2'-methoxyacetophenone 98%
Irritant
4'-Fluoro-2'-methoxyacetophenone 98%
Catalog # APOSPC3817C-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4'-Fluoro-2'-methoxyacetophenone 98%
Catalog # APOSPC3817C-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :168.17 g/mol
Melting Pt:51…53 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:51788-80-8
Formula:C₉H₉FO₂
MDL Number:MFCD00061143