Order Entry
Germany
Orders LinkContactUsLinkComponent
3-Fluoropyrocatechol 99%

3-Fluoropyrocatechol 99%

Corrosive
Irritant
Supplier:   Apollo Scientific
3-Fluoropyrocatechol 99%
Supplier:  Apollo Scientific
Certificates
3-Fluoropyrocatechol 99%

3 Options Available Below

Product Details & Documents
Synonyms:  3-Fluorocatechol, 3-Fluorobenzene-1,2-diol, 3-Fluoro-1,2-dihydroxybenzene
  • CAS Number:363-52-0
  • Formula:C₆H₅FO₂
  • MDL Number:MFCD00042582
  • Molecular Weight:128.1 g/mol
  • Melting Pt:71…73 °C
  • Storage Temperature:Ambient
  • UN:3261
  • ADR:8,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 39,10 €
    39,10 €
    1 * 5 g
       39,10 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 132,00 €
    132,00 €
    1 * 25 g
       132,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.