Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Fluorobenzyl mercaptan
4-Fluorobenzyl mercaptan 98%
4-Fluorobenzyl mercaptan 98%
:
Apollo Scientific
4-Fluorobenzyl mercaptan 98%
:
Apollo Scientific
:
15894-04-9
:
:
C₇H₇FS
:
MFCD00040724
:
142.2 g/mol
:
72…74 °C/15mmHg
:
1.157 g/cm³ (25 °C)
:
76 °C
:
Refrigerator
:
3334
:
9,III
:
GHS07