Order Entry
Germany
Orders LinkContactUsLinkComponent
 

2'-Fluoroacetophenone 97%

Irritant
   Sustainable Options Available
Supplier:   Apollo Scientific
2'-Fluoroacetophenone 97%
Supplier:  Apollo Scientific
   Sustainable Options Available
Certificates

2 Options Available Below

Product Details & Documents

Reaction of o-fluoroketones with guanidine carbonate affords 2-amino-4-alkyl- & 2- amino-4-arylquinazolines.

Synonyms:  1-(2-Fluorophenyl)ethanone, o-Fluoroacetophenone
  • CAS Number:445-27-2
  • Formula:C₈H₇FO
  • MDL Number:MFCD00000320
  • Molecular Weight:138.14 g/mol
  • Boiling Pt:187…189 °C
  • Density:1.137 g/cm³ (25 °C)
  • Flash Pt:61 °C
  • Storage Temperature:Ambient
  • EINECS:207-156-0
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 21,80 €
    21,80 €
    1 * 25 g
       21,80 €

    Specifications:
    More
    Specifications
    Cat. No.
    APOSPC3360-25G
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 39,10 €
    39,10 €
    1 * 100 g
       39,10 €

    Specifications:
      
    More
    Specifications
    Cat. No.
    APOSPC3360-100G
      
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.