3-Bromo-2-(tert-butyl)-7-chloro-5-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:655235-50-0
:
Formula:C₁₁H₁₀BrClF₃N₃
MDL Number:MFCD00178915
:356.58 g/mol
Storage Temperature:Ambient