Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Ethoxy-2-(trifluoromethyl)phenylboronic acid
4-Ethoxy-2-(trifluoromethyl)phenylboronic acid 98%
4-Ethoxy-2-(trifluoromethyl)phenylboronic acid 98%
Irritant
4-Ethoxy-2-(trifluoromethyl)phenylboronic acid 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Ethoxy-2-(trifluoromethyl)phenylboronic acid 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
313545-39-0
:
Formula:
C₉H₁₀BF₃O₃
MDL Number:
MFCD08059515
:
233.98 g/mol
:
GHS07
Storage Temperature:
Refrigerator